missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-2,3-dihydro-1-benzofuran-7-amine, 95%, Thermo Scientific™

Product Code. 10183214
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10183214 1 g Amber Glass Bottle 1g
10123174 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10183214 Supplier Thermo Scientific Maybridge Supplier No. CC27214DA
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 68298-46-4
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD00075581
InChI Key LMTIGABGABPAGU-UHFFFAOYSA-N
Synonym 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-amine, unii-c3e03fdp2f, 2,3-dihydro-2,2-dimethyl-7-aminobenzofuran, 7-amino-2,3-dihydro-2,2-dimethylbenzofuran, 7-benzofuranamine, 2,3-dihydro-2,2-dimethyl, c3e03fdp2f, 2,3-dihydro-2,2-dimethylbenzofuran-7-amine, 2,2-dimethyl-2,3-dihydrobenzofuran-7-amine, 2,3-dihydro-2,2-dimethyl-7-benzofuranamine, dsstox_cid_24715
PubChem CID 91697
SMILES CC1(C)CC2=CC=CC(N)=C2O1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-amine
Formula Weight 163.22
Percent Purity 95%
Chemical Name or Material 2, 2-Dimethyl-2, 3-dihydro-1-benzofuran-7-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.