missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-3,4-dihydro-2H-1,4-benzoxazine, 97%

Product Code. 15406236
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15406236 250 mg 250mg
15497548 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15406236 Supplier Thermo Scientific Alfa Aesar Supplier No. H61058.MD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 866089-28-3 | C10H13NO | 163.22 g/mol

It possess various biological activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It possess various biological activities.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 866089-28-3
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD11603419
InChI Key WILBVUMFTFLAHJ-UHFFFAOYSA-N
Synonym 2,2-dimethyl-3,4-dihydro-2h-1,4-benzoxazine, 2,2-dimethyl-3,4-dihydro-2h-benzo b 1,4 oxazine, 2h-1,4-benzoxazine,3,4-dihydro-2,2-dimethyl, 2,2-dimethyl-3,4-dihydro-2h-benzo 1,4 oxazine, 2h-1,4-benzoxazine, 3,4-dihydro-2,2-dimethyl
PubChem CID 23587101
IUPAC Name 2,2-dimethyl-3,4-dihydro-1,4-benzoxazine
SMILES CC1(CNC2=CC=CC=C2O1)C

Specifications

Quantity 250 mg
Formula Weight 163.22
Percent Purity 97%
Chemical Name or Material 2,2-Dimethyl-3,4-dihydro-2H-1,4-benzoxazine
compliance-icons
Product Identifier
  • 2,2-Dimethyl-3,4-dihydro-2H-1,4-benzoxazine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2,2-Dimethyl-3,4-dihydro-2H-1,4-benzoxazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.