missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-N-(4-pyridinyl)propanamide, 97%, Thermo Scientific™

Product Code. 10364482
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10364482 1 g Amber Glass Bottle 1g
10601113 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10364482 Supplier Thermo Scientific Maybridge Supplier No. MO00094DA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 70298-89-4
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
MDL Number MFCD00996248
InChI Key JCMMVFHXRDNILC-UHFFFAOYSA-N
Synonym 2,2-dimethyl-n-pyridin-4-yl-propionamide, n-pyridin-4-yl pivalamide, 2,2-dimethyl-n-pyridin-4-yl propanamide, 2,2-dimethyl-n-4-pyridinyl propanamide, n-4-pyridyl pivalamide, 2,2-dimethyl-n-4-pyridyl propanamide, 4-2,2,2-trimethylacetamido pyridine, 2,2-dimethyl-n-4-pyridyl propionamide, 4-pivaloylaminopyridine, maybridge3_004407
PubChem CID 427059
IUPAC Name 2,2-dimethyl-N-pyridin-4-ylpropanamide
SMILES CC(C)(C)C(=O)NC1=CC=NC=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 178.23
Percent Purity 97%
Chemical Name or Material 2, 2-Dimethyl-N-(4-pyridinyl)propanamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.