missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(Dimethylamino)ethoxy]aniline, 97%, Thermo Scientific™

Product Code. 10401514
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
10401514 250 mg 250mg
10340274 1 g 1g
10336954 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 10401514

Brand: Thermo Scientific Maybridge CC51314CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1202-00-2
Molecular Formula C10H16N2O
Molecular Weight (g/mol) 180.251
InChI Key OHDPNHWSANKRNF-UHFFFAOYSA-N
Synonym 2-2-dimethylamino ethoxy aniline, 2-2-dimethylamino-ethoxy-phenylamine, benzenamine,2-2-dimethylamino ethoxy, benzenamine, 2-2-dimethylamino ethoxy, 2-2-aminophenoxy ethyl dimethylamine, 2-2-dimethylaminoethyloxy aniline
PubChem CID 6484656
IUPAC Name 2-[2-(dimethylamino)ethoxy]aniline
SMILES CN(C)CCOC1=CC=CC=C1N

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 180.25
Percent Purity 97%
Chemical Name or Material 2-[2-(Dimethylamino)ethoxy]aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.