missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylbutane 98%, Thermo Scientific™

Product Code. 10509420 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10509420 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10509420 Supplier Thermo Scientific Acros Supplier No. 116091000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-83-2
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD00009321
InChI Key HNRMPXKDFBEGFZ-UHFFFAOYSA-N
Synonym neohexane, butane, 2,2-dimethyl, hsdb 75, unii-07l56l3mp2, ccris 6019, 2,2-dimethylbutyl, 3,3-dimethylbutyl, butane,2-dimethyl, 2,2-dimethyl-butane, 3,3-dimethylbutan-2-yl
PubChem CID 6403
IUPAC Name 2,2-dimethylbutane
SMILES CCC(C)(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.6490g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C2H5C(CH3)3
Refractive Index 1.3675 to 1.3695 (20°C, 589nm)
Quantity 100 mL
Beilstein 01, 150
Formula Weight 86.18
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 2, 2-Dimethylbutane, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.