missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylbutanoic Acid, TRC

Product Code. 30387021
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30387021 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387021 Supplier Toronto Research Chemicals Supplier No. TRCD46804025G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2-Dimethylbutanoic Acid
CAS 595-37-9
Molecular Formula C6H12O2
Quantity 25 g
Synonym 2,2-Dimethylbutyric Acid, Neohexanoic Acid, alpha,alpha-Dimethylbutanoic Acid, alpha,alpha-Dimethylbutyric Acid, NSC 16045, NSC 741804
InChI Formula InChI=1S/C6H12O2/c1-4-6(2,3)5(7)8/h4H2,1-3H3,(H,7,8)
SMILES CCC(C)(C)C(=O)O
IUPAC Name 2,2-dimethylbutanoic acid
Molecular Weight (g/mol) 116.16
Formula Weight 116.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.