missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylsuccinic acid, 99%

Product Code. 11455837
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455837 5 g 5g
11445837 1 g 1g
11465837 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455837 Supplier Thermo Scientific Alfa Aesar Supplier No. B24394.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 597-43-3 | C6H10O4 | 146.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 597-43-3
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.14
MDL Number MFCD00004196
InChI Key GOHPTLYPQCTZSE-UHFFFAOYSA-N
Synonym 2,2-dimethylsuccinic acid, 2,2-dimethyl-succinic acid, unii-mx6tvr2107, 2,2-dimethyl-butanedioic acid, succinic acid, 2,2-dimethyl, butanedioic acid, 2,2-dimethyl, 2,2-dimethylbutanedioate, 2,2-dimethyl-butanedioicacid, 2,2,-dimethylsuccinate, 2,2-dimethyl succinate
PubChem CID 11701
ChEBI CHEBI:86537
IUPAC Name 2,2-dimethylbutanedioic acid
SMILES CC(C)(CC(O)=O)C(O)=O

Specifications

Melting Point 139°C to 142°C
Quantity 5 g
Beilstein 1764476
Formula Weight 146.14
Percent Purity 99%
Chemical Name or Material 2,2-Dimethylsuccinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.