missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dipyridinyl Disulfide, TRC

Product Code. 30369465
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30369465 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30369465 Supplier Toronto Research Chemicals Supplier No. TRCD49275025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2'-Dipyridinyl Disulfide
CAS 2127-03-9
Molecular Formula C10 H8 N2 S2
Quantity 25 g
Synonym 2,2'-Dithiobis-pyridine, 1,2-Bis(2-pyridinyl) Disulfide, 2,2'-Bipyridyldisulfide, 2,2'-Dithiobis(pyridine), 2,2'-Dithiodipyridine, 2-Aldrithiol, Di-2-pyridyl Disulfide, NSC 677438, NSC 94055
InChI Formula InChI=1S/C10H8N2S2/c1-3-7-11-9(5-1)13-14-10-6-2-4-8-12-10/h1-8H
SMILES S(Sc1ccccn1)c2ccccn2
IUPAC Name 2-(pyridin-2-yldisulfanyl)pyridine
Molecular Weight (g/mol) 220.31
Formula Weight 220.0129
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.