missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dipyridyl, 99.5+%, Certified AR for Analysis, Fisher Chemical

Product Code. 10449170 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Plastic powder jar
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10449170 5 g Plastic powder jar 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10449170 Supplier Fisher Chemical Supplier No. D/5051/44

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 366-18-7
Melting Point 69°C
Color White
pH 7.5
Boiling Point 272°C
Packaging Plastic powder jar
Molecular Formula C10H8N2
MDL Number MFCD00006212
Quantity 5 g
Synonym 2,2'-bipyridine, 2,2'-dipyridyl, 2,2'-bipyridyl, bipyridine, 2,2'-dipyridine, 2-2-pyridyl pyridine, 2,2'-bipyridin, alpha,alpha'-dipyridyl, alpha,alpha'-bipyridyl, 2,2-bipyridyl
InChI Key ROFVEXUMMXZLPA-UHFFFAOYSA-N
SMILES C1=CC=C(N=C1)C1=CC=CC=N1
IUPAC Name 2,2'-bipyridine
Molecular Weight (g/mol) 156.19
PubChem CID 1474
ChEBI CHEBI:30351
Formula Weight 156.19g/mol
Physical Form Powder
Show More Show Less
compliance-icons
Product Identifier
  • 2,2``-Dipyridyl
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.