missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dipyridyl, 99+%

Product Code. 10080810 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10080810 10 g Glass bottle 10g
10274570 25 g Glass bottle 25g
10530581 100 g Glass bottle 100g
11936291 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10080810 Supplier Thermo Scientific Acros Supplier No. 117500100
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 366-18-7 | C10H8N2 | 156.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 366-18-7
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD00006212
InChI Key ROFVEXUMMXZLPA-UHFFFAOYSA-N
Synonym 2,2'-bipyridine, 2,2'-dipyridyl, 2,2'-bipyridyl, bipyridine, 2,2'-dipyridine, 2-2-pyridyl pyridine, 2,2'-bipyridin, alpha,alpha'-dipyridyl, alpha,alpha'-bipyridyl, 2,2-bipyridyl
PubChem CID 1474
ChEBI CHEBI:30351
IUPAC Name 2-pyridin-2-ylpyridine
SMILES C1=CC=C(N=C1)C1=CC=CC=N1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 69.0°C to 72.0°C
Color White
Boiling Point 272.0°C to 273.0°C
Flash Point 121°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 23,199
Fieser 16,25
Merck Index 15,1241
Solubility Information Solubility in water: slightly soluble (5.5g/L 22°C). Other solubilities: very soluble in ethanol (500g/L 25°C), ether,, benzene, chloroform and petroleum ether
Viscosity 1.51 mm2/s (90°C)
Formula Weight 156.19
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 2, 2'-Dipyridyl
Show More Show Less
compliance-icons
Product Identifier
  • 2,2``-Dipyridyl
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 2,2-Bipyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.