missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dithiobisbenzanilide, TRC

Product Code. 30369589
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30369589 2.5 g
30376033 250 mg
30385521 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30369589 Supplier Toronto Research Chemicals Supplier No. TRCD4942132.5G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-57-9
Molecular Formula C26H20N2O2S2
Molecular Weight (g/mol) 456.58
Synonym 2,2'-Dibenzoylaminodiphenyldisulfid, Benzamide, N,N'-(dithiodi-2,1-phenylene)bis-, Benzanilide, 2',2'''-dithiobis- (6CI,7CI,8CI), N,N'-(Dithiodi-2,1-phenylene)bis[benzamide], 2,2'-Dibenzamidodiphenyl disulfide, 2,2'-Dibenzoylaminodiphenyl disulfide, 2',2'''-Dithiodibenzanilide, 3S, Bis(2-benzamidophenyl) disulfide, Bis(2-benzoylaminophenyl)disulfide, Bis(o-benzamidophenyl) disulfide, Bisbenzamide, DBD, Di-o-benzamidophenyl disulfide, K 202, Noctizer SS, Peptazin BAFD, Peptisant 10, Pepton 22, Pepton 3S, Renazit 11, SL 6010, SS, Vuchtazin RV/G-M, Vuchtazin RV/G-S, o,o'-Dibenzamidodiphenyl disulfide, o-(Benzoylamino)phenyl disulfide
SMILES O=C(Nc1ccccc1SSc2ccccc2NC(=O)c3ccccc3)c4ccccc4

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C26H20N2O2S2/c29-25(19-11-3-1-4-12-19)27-21-15-7-9-17-23(21)31-32-24-18-10-8-16-22(24)28-26(30)20-13-5-2-6-14-20/h1-18H,(H,27,29)(H,28,30)
Formula Weight 456.0966
Chemical Name or Material 2,2'-Dithiobisbenzanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.