missing translation for 'onlineSavingsMsg'
Learn More

2(2-Ethoxyethoxy)ethanol, 98+%

Product Code. 10793651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
Unit Size:
1 L
2.5 L
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10793651 1 L Glass Bottle 1 L
10182810 2.5 L Glass Bottle 2.5 L
10016253 25 L Metal drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10793651 Supplier Thermo Scientific Acros Supplier No. 117890010
  • ONLINE EXCLUSIVE PRICE
    50.40 €
    48.90 € / 1 L
    Save 1.50 €
    2% Off
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 111-90-0 | C6H14O3 | 134.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-90-0
MDL Number MFCD00002872
InChI Key XXJWXESWEXIICW-UHFFFAOYSA-N
Synonym diethylene glycol monoethyl ether, 2-2-ethoxyethoxy ethanol, carbitol, transcutol, ethoxy diglycol, dioxitol, ethyl carbitol, ethyl digol, carbitol solvent, ethanol, 2-2-ethoxyethoxy
PubChem CID 8146
ChEBI CHEBI:40572
IUPAC Name 2-(2-ethoxyethoxy)ethanol
SMILES CCOCCOCCO

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -80.0°C
Color Colorless
Density 0.9900g/mL
Boiling Point 197.0°C
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass Bottle
Molecular Formula C6H14O3
Linear Formula C2H5OCH2CH2OCH2CH2OH
Refractive Index 1.4260 to 1.4280
Quantity 1 L
Beilstein 01, II, 520
Merck Index 15, 3134
Specific Gravity 0.99
Molecular Weight (g/mol) 134.175
Formula Weight 134.17
Percent Purity ≥98%
Grade Pure
Physical Form Liquid
Chemical Name or Material 2(2-Ethoxyethoxy)ethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.