missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Ethoxyphenoxy)ethyl bromide, 98%, Thermo Scientific™

Product Code. 11359156
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359156 5 g 5g
11369156 25 g 25g
11379156 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11359156 Supplier Thermo Scientific Alfa Aesar Supplier No. L20300.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3-8-3259
MDL Number MFCD02030483

Specifications

Melting Point 41°C to 43°C
Molecular Formula C10H13BrO2
Quantity 5 g
Beilstein 6290077
Synonym 2-2-ethoxyphenoxy ethyl bromide, 1-2-bromoethoxy-2-ethoxybenzene, 2-2-ethoxyphenoxy ethylbromide, 2-2-bromoethoxy phenetole, benzene, 1-2-bromoethoxy-2-ethoxy, 1-2-bromoethoxy-2-ethyoxybenzene, 1-2-bromoethoxy-2-ethoxy-benzene, 2-bromo-1-2-ethoxyphenoxy ethane, tamsulosin impurity i, pubchem13149
InChI Key IOYHGBZPUZBUTJ-UHFFFAOYSA-N
SMILES CCOC1=CC=CC=C1OCCBr
IUPAC Name 1-(2-bromoethoxy)-2-ethoxybenzene
Molecular Weight (g/mol) 245.12
PubChem CID 7010183
Formula Weight 245.12
Percent Purity 98%
Chemical Name or Material 2-(2-Ethoxyphenoxy)ethyl bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.