missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Ethoxyphenoxy)ethyl bromide, 98%, Thermo Scientific™

Product Code. 11379156
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11379156 100 g 100g
11359156 5 g 5g
11369156 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11379156 Supplier Thermo Scientific Alfa Aesar Supplier No. L20300.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3-8-3259
MDL Number MFCD02030483

Specifications

Melting Point 41°C to 43°C
Molecular Formula C18H12N2O4
Quantity 100 g
Beilstein 6290077
Synonym 4,4-dinitro-p-terphenyl, p-terphenyl, 4,4-dinitro, 4,4-dinitro-1,1',4',1 terphenyl, 1,4-bis 4-nitrophenyl benzene, p,p-dinitro-p-terphenyl, 4,4-dinitro-para-terphenyl, 4,4-dinitro-1,1',4',1-terphenyl, 4,4-dinitro-1,1':4',1-terphenyl, 1-nitro-4-4-4-nitrophenyl phenyl benzene, 4,4-dinitro-1,1':4',1-terbenzene
InChI Key MHOAYDHUNGLDTB-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)[N+](=O)[O-])C3=CC=C(C=C3)[N+](=O)[O-]
IUPAC Name 1,4-bis(4-nitrophenyl)benzene
Molecular Weight (g/mol) 320.30
PubChem CID 101423
Formula Weight 245.12
Percent Purity 98%
Chemical Name or Material 2-(2-Ethoxyphenoxy)ethyl bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.