missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Formamidothiazol-4-yl)acetic Acid-13C,15N2, TRC

Product Code. 30398452
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398452 25 mg 25mg
30392065 5 mg 5mg
30398920 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30398452 Supplier Toronto Research Chemicals Supplier No. TRCF69211725MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(2-Formamidothiazol-4-yl)acetic Acid-13C,15N2
Molecular Formula 13C C5 H6 15N2 O3 S
Quantity 25 mg
InChI Formula InChI=1S/C6H6N2O3S/c9-3-7-6-8-4(2-12-6)1-5(10)11/h2-3H,1H2,(H,10,11)(H,7,8,9)/i6+1,7+1,8+1
SMILES OC(=O)Cc1cs[13c]([15NH]C=O)[15n]1
IUPAC Name 2-[2-(formyl(15N)amino)-(213C,315N)1,3-thiazol-4-yl]acetic acid
Molecular Weight (g/mol) 189.168
Formula Weight 189.007
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.