missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Furyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10694585
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10694585 5 g Amber Glass Bottle 5g
10507284 250 mg Amber Glass Bottle 250mg
10336904 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10694585 Supplier Thermo Scientific Maybridge Supplier No. CC46201DE
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 331942-47-3
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.182
InChI Key QRUHYAWZHFTNEA-UHFFFAOYSA-N
Synonym 2-2-furyl benzoic acid, 2-furan-2-yl benzoic acid, 2-fur-2-ylbenzoic acid, benzoic acid,2-2-furanyl, 2-fur-2-yl benzoic acid, 2-furan-2-yl-benzoic acid
PubChem CID 2772293
IUPAC Name 2-(furan-2-yl)benzoic acid
SMILES C1=CC=C(C(=C1)C2=CC=CO2)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 188.18
Percent Purity 97%
Chemical Name or Material 2-(2-Furyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.