missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Furyl)benzonitrile 97+%, Thermo Scientific™

Product Code. 10255914
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Product Code. Quantity Packaging unitSize
10255914 250 mg Amber Glass Bottle 250mg
10757285 1 g Amber Glass Bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10255914

Brand: Thermo Scientific Maybridge CC46216CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 155395-45-2
Molecular Formula C11H7NO
Molecular Weight (g/mol) 169.18
MDL Number MFCD04039074
InChI Key XLZNEBADSPRURH-UHFFFAOYSA-N
Synonym 2-2-furyl benzonitrile, 2-furan-2-yl benzonitrile, 2-furan-2-yl-benzonitrile, benzonitrile,2-2-furanyl, 2-2-furanyl benzonitrile, 2-2-furyl benzenecarbonitrile, 2-furan-2-yl benzenecarbonitrile
PubChem CID 4335608
IUPAC Name 2-(furan-2-yl)benzonitrile
SMILES N#CC1=CC=CC=C1C1=CC=CO1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 169.18
Percent Purity 97%
Chemical Name or Material 2-(2-Furyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.