missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Furyl)benzonitrile 97+%, Thermo Scientific™

Product Code. 10757285
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10757285 1 g Amber Glass Bottle 1g
10255914 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10757285 Supplier Thermo Scientific Maybridge Supplier No. CC46216DA
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 155395-45-2
Molecular Formula C11H7NO
Molecular Weight (g/mol) 169.18
MDL Number MFCD04039074
InChI Key XLZNEBADSPRURH-UHFFFAOYSA-N
Synonym 2-2-furyl benzonitrile, 2-furan-2-yl benzonitrile, 2-furan-2-yl-benzonitrile, benzonitrile,2-2-furanyl, 2-2-furanyl benzonitrile, 2-2-furyl benzenecarbonitrile, 2-furan-2-yl benzenecarbonitrile
PubChem CID 4335608
IUPAC Name 2-(furan-2-yl)benzonitrile
SMILES N#CC1=CC=CC=C1C1=CC=CO1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 169.18
Percent Purity 97%
Chemical Name or Material 2-(2-Furyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.