missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Hydroxyethyl)pyridine, 98%, Thermo Scientific Chemicals

Product Code. 11495607
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495607 500 g 500g
11485607 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11495607 Supplier Thermo Scientific Alfa Aesar Supplier No. B23579.36
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 103-74-2 | Molecular Formula: C7H9NO | Molecular Weight: 123.155 g/mol

2-(2-Hydroxyethyl)pyridine is a metabolite of the anti-vertigo drug Betahistine. It acts as a reagent for protection of the 5'-phosphate of nucleotides as 2-pyridylethyl (Pet) esters. It is used for carboxyl protection of amino acids. It is also used to cleave such groups as Boc and Fmoc, and also to hydrogenolysis. It was used to prepare mononuclear nickel(II) acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Hydroxyethyl)pyridine is a metabolite of the anti-vertigo drug Betahistine. It acts as a reagent for protection of the 5′-phosphate of nucleotides as 2-pyridylethyl (Pet) esters. It is used for carboxyl protection of amino acids. It is also used to cleave such groups as Boc and Fmoc, and also to hydrogenolysis. It was used to prepare mononuclear nickel(II) acetate.

Solubility
Soluble in Chloroform (Slightly), Methanol (Slightly).

Notes
Hygroscopic. Store under dry inert gas. Store away from moisture, strong bases and oxidizing agents. Protect from humidity and water.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-74-2
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00006364
InChI Key BXGYBSJAZFGIPX-UHFFFAOYSA-N
Synonym 2-pyridineethanol, 2-2-hydroxyethyl pyridine, 2-2-pyridyl ethanol, pyridine-2-ethanol, 2-pyridin-2-yl ethan-1-ol, 2-pyridin-2-yl ethanol, 2-pyridinethanol, 2-pyridylethanol, 2-pyridine ethanol, 2-beta-hydroxyethyl pyridine
PubChem CID 7675
IUPAC Name 2-pyridin-2-ylethanol
SMILES C1=CC=NC(=C1)CCO

Spécification

Melting Point -8°C
Density 1.095
Boiling Point 235°C to 238°C
Flash Point 117°C (243°F)
Refractive Index 1.537
Quantity 500 g
Beilstein 111205
Sensitivity Hygroscopic
Solubility Information Soluble in Chloroform (Slightly),Methanol (Slightly).
Formula Weight 123.16
Percent Purity 99%
Chemical Name or Material 2-(2-Hydroxyethyl)pyridine
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.