missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methoxyethoxy)ethanol, 99%

Product Code. 11984731 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11984731 10 L Plastic drum 10 L
10041590 1 L Glass Bottle 1 L
10051590 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11984731 Supplier Thermo Scientific Acros Supplier No. 149370100
  • ONLINE EXCLUSIVE PRICE
    244.00 €
    223.00 € / 10 L
    Save 21.00 €
    8% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 111-77-3 | C5H12O3 | 120.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-77-3
Molecular Formula C5H12O3
Molecular Weight (g/mol) 120.15
MDL Number MFCD00002871,MFCD00084416
InChI Key SBASXUCJHJRPEV-UHFFFAOYSA-N
Synonym 2-2-methoxyethoxy ethanol, diethylene glycol monomethyl ether, methyl carbitol, methyl digol, methoxydiglycol, ethanol, 2-2-methoxyethoxy, methyl dioxitol, diethylene glycol methyl ether, dowanol dm, poly-solv dm
PubChem CID 8134
ChEBI CHEBI:44836
IUPAC Name 2-(2-methoxyethoxy)ethanol
SMILES COCCOCCO

Specifications

Melting Point -70.0°C
Density 1.0100g/mL
Boiling Point 194.0°C
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic drum
Linear Formula CH3OCH2CH2OCH2CH2OH
Refractive Index 1.4250 to 1.4270
Quantity 10 L
Beilstein 01,IV,2392
Merck Index 15,3136
Specific Gravity 1.01
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol,glycerol,ether,acetone,and dmf
Viscosity 3.9 mPa.s (20°C)
Formula Weight 120.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-(2-Methoxyethoxy)ethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.