missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methoxyphenoxy)ethylamine Hydrochloride, TRC

Product Code. 30012087
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012087 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30012087 Supplier Toronto Research Chemicals Supplier No. TRCM2656501G
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 64464-07-9
Molecular Formula C9 H13 N O2 . Cl H
Quantity 1 g
Synonym 2-(2-Methoxyphenoxy)-ethanamine hydrochloride, 2-(o-Methoxyphenoxy)-ethylamine hydrochloride, 2-(2-Methoxyphenoxy)ethylamine Hydrochloride
InChI Formula InChI=1S/C9H13NO2.ClH/c1-11-8-4-2-3-5-9(8)12-7-6-10;/h2-5H,6-7,10H2,1H3;1H
SMILES Cl.COc1ccccc1OCCN
IUPAC Name 2-(2-methoxyphenoxy)ethanamine;hydrochloride
Molecular Weight (g/mol) 203.67
Formula Weight 203.0713
Chemical Name or Material 2-(2-Methoxyphenoxy)ethylamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.