missing translation for 'onlineSavingsMsg'
Learn More

2-[(2-Methoxyphenoxy)methyl]oxirane, TRC

Product Code. 30477024
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477024 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477024 Supplier Toronto Research Chemicals Supplier No. TRCM265700500MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[(2-Methoxyphenoxy)methyl]oxirane
CAS 2210-74-4
Molecular Formula C10 H12 O3
Quantity 500 mg
Synonym 2-[(2-Methoxyphenoxy)methyl]oxirane, Oxirane','2-[(2-methoxyphenoxy)methyl]-, Benzene','1-(2,3-epoxypropoxy)-2-methoxy- (6CI,7CI,8CI), Oxirane','[(2-methoxyphenoxy)methyl]- (9CI), 1,2-Epoxy-3-(2-methoxyphenoxy)propane, 1,2-Epoxy-3-(o-methoxyphenoxy)propane, 1-(2,3-Epoxypropoxy)-2-methoxybenzene, 1-(2-Methoxyphenoxy)-2,3-epoxypropane, 2-(2,3-Epoxypropoxy)anisole, 2-Methoxy-1-(oxiranylmethoxy)benzene, 2-Methoxyphenyl glycidyl ether, Glycidyl 2-methoxyphenyl ether, Glycidyl o-methoxyphenyl ether, NSC 112256, NSC 133442
InChI Formula InChI=1S/C10H12O3/c1-11-9-4-2-3-5-10(9)13-7-8-6-12-8/h2-5,8H,6-7H2,1H3
SMILES COc1ccccc1OCC2CO2
IUPAC Name 2-[(2-methoxyphenoxy)methyl]oxirane
Molecular Weight (g/mol) 180.2
Formula Weight 180.0786
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.