missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methoxyphenyl)piperazine monohydrate, 95%

Product Code. 15487575
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487575 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15487575 Supplier Thermo Scientific Alfa Aesar Supplier No. H26408.MD
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 65709-27-5 | C11H16N2O | 192.262 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 65709-27-5
Melting Point 77°C to 83°C
Molecular Formula C11H16N2O
MDL Number MFCD06660448
Quantity 250 mg
UN Number UN3259
Sensitivity Air Sensitive
Synonym 2-2-methoxyphenyl piperazine, 2-2-methoxyphenyl-piperazine, 2-2-methoxy-phenyl-piperazine, 2-methoxyphenyl piperazine, acmc-1bd94, 1-methoxy-2-piperazin-2-ylbenzene
InChI Key XYMDIQVPOIVJPF-UHFFFAOYSA-N
SMILES COC1=CC=CC=C1C2CNCCN2
IUPAC Name 2-(2-methoxyphenyl)piperazine
Molecular Weight (g/mol) 192.262
PubChem CID 16740792
Formula Weight 210.28 (192.26 Anhydrous)
Percent Purity 95%
Chemical Name or Material 2-(2-Methoxyphenyl)piperazine monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.