missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methyl-1,3-thiazol-4-yl)ethylamine, 97%, Thermo Scientific™

Product Code. 10122844
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10122844 250 mg Amber Glass Bottle 250mg
10671934 1 g Amber Glass Bottle 1g
10737485 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10122844 Supplier Thermo Scientific Maybridge Supplier No. CC54613CB
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 165115-15-1
Molecular Formula C6H10N2S
Molecular Weight (g/mol) 142.22
MDL Number MFCD03011601
InChI Key GLEZGSKBHYTSAD-UHFFFAOYSA-N
Synonym 2-2-methyl-1,3-thiazol-4-yl ethanamine, 2-2-methylthiazol-4-yl ethanamine, 2-2-methyl-1,3-thiazol-4-yl ethylamine, 4-thiazoleethanamine, 2-methyl, 2-2-methyl-1,3-thiazol-4-yl ethan-1-amine, 4-thiazoleethanamine,2-methyl, 2-2-methyl-1,3-thiazol-4-yl-1-ethanamine
PubChem CID 7536518
IUPAC Name 2-(2-methyl-1,3-thiazol-4-yl)ethanamine
SMILES CC1=NC(CCN)=CS1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 142.23
Percent Purity 97%
Chemical Name or Material 2-(2-Methyl-1, 3-thiazol-4-yl)ethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.