missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methyl-1H-imidazol-1-yl)benzonitrile, 97%, Thermo Scientific™

Product Code. 10748635
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10748635 5 g Amber Glass Bottle 5g
10707295 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10748635 Supplier Thermo Scientific Maybridge Supplier No. CC61216DE
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 892502-27-1
Molecular Formula C11H9N3
Molecular Weight (g/mol) 183.214
MDL Number MFCD09065013
InChI Key NCQQCFZREDABTC-UHFFFAOYSA-N
Synonym 2-2-methyl-1h-imidazol-1-yl benzonitrile, 2-2-methylimidazol-1-yl benzonitrile, 2-2-methylimidazolyl benzenecarbonitrile, benzonitrile,2-2-methyl-1h-imidazol-1-yl
PubChem CID 24229637
IUPAC Name 2-(2-methylimidazol-1-yl)benzonitrile
SMILES CC1=NC=CN1C2=CC=CC=C2C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 183.21
Percent Purity 97%
Chemical Name or Material 2-(2-Methyl-1H-imidazol-1-yl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.