missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Phthalimidoethoxy)acetic acid, 97%

Product Code. 15466887
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15466887 1 g 1g
15476887 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15466887 Supplier Thermo Scientific Alfa Aesar Supplier No. H33988.03
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 69676-65-9 | C12H11NO5 | 249.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69676-65-9
Molecular Formula C12H11NO5
Molecular Weight (g/mol) 249.22
MDL Number MFCD06661364
InChI Key PHZYUQLIZKTSJE-UHFFFAOYSA-N
Synonym 2-2-1,3-dioxoisoindolin-2-yl ethoxy acetic acid, 2-2-phthalimidoethoxy acetic acid, 2-2-phthalimidoethoxy-acetic acid, 2-1,3-dioxoisoindol-2-yl ethoxy acetic acid, 2-2-1,3-dioxoisoindol-2-yl ethoxy acetic acid, 2-phthalimido ethoxyacetic acid, 2-1,3-dioxoisoindolin-2-yl ethoxy acetic acid, 2-2-1,3-dioxoisoindolin-2-yl ethoxy aceticacid, 2-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl ethoxy acetic acid, 2-2-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl ethoxy acetic acid
PubChem CID 2755407
IUPAC Name 2-[2-(1,3-dioxoisoindol-2-yl)ethoxy]acetic acid
SMILES OC(=O)COCCN1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 130°C to 132°C
Quantity 1 g
Formula Weight 249.22
Percent Purity 97%
Chemical Name or Material 2-(2-Phthalimidoethoxy)acetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.