missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Propynyloxy)tetrahydro-2H-pyran, 98%

Product Code. 10617694 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10617694 5 g Glass bottle 5g
10279363 25 g Glass bottle 25g
10569453 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10617694 Supplier Thermo Scientific Acros Supplier No. 295060050
 Restricted Item
Estimated Shipment: 17-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6089-04-9 | C8H12O2 | 140.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6089-04-9
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00006604
InChI Key HQAXHIGPGBPPFU-UHFFFAOYNA-N
Synonym tetrahydro-2-2-propynyloxy-2h-pyran, 2-2-propynyloxy tetrahydropyran, 2-propargyloxane, 2-prop-2-yn-1-yloxy tetrahydro-2h-pyran, 2-propargyloxy tetrahydropyran, propargyl 2-tetrahydropyranyl ether, propargyl alcohol tetrahydropyranyl ether, 2h-pyran, tetrahydro-2-2-propynyloxy, 2-propynyl 2-pyranyl ether, 3-2'-tetrahydropyranyloxy propyne
PubChem CID 98609

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9970g/mL
Boiling Point 63°C to 65°C (9.0 mmHg)
Flash Point 59°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4560 to 1.458
Quantity 5 g
Specific Gravity 0.997
Formula Weight 140.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-(2-Propynyloxy)tetrahydro-2H-pyran
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-(2-Propynyloxy)- tetrahydro-2H-pyran
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.