missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Pyrrolidinyl)pyridine, 96%

Product Code. 11459290
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459290 250 mg 250mg
11469290 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459290 Supplier Thermo Scientific Alfa Aesar Supplier No. H50176.MD
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 77790-61-5 | C9H12N2 | 148.209 g/mol

2-(2-Pyrrolidinyl)pyridine is used as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Pyrrolidinyl)pyridine is used as primary and secondary intermediate.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77790-61-5
Molecular Formula C9H12N2
Molecular Weight (g/mol) 148.209
MDL Number MFCD01691538
InChI Key NDCZQFDBSPOUDF-UHFFFAOYSA-N
Synonym 2-pyrrolidin-2-yl pyridine, 2-2-pyrrolidinyl pyridine, 2-pyrrolidin-2-yl-pyridine, +--alpha-nornicotine, pyridine, 2-2-pyrrolidinyl, +--2-2-pyrrolidinyl pyridine, pyridine, 2-2-pyrrolidinyl-, +-, r-2-2-pyrrolidinyl-pyridine, alpha-nornicotine, 2,3'-pyridylpyrrolidine
PubChem CID 2771659
IUPAC Name 2-pyrrolidin-2-ylpyridine
SMILES C1CC(NC1)C2=CC=CC=N2

Specifications

Quantity 250 mg
UN Number UN2810
Solubility Information Soluble in water.
Formula Weight 148.21
Percent Purity 96%
Chemical Name or Material 2-(2-Pyrrolidinyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.