missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Thienyl)ethyl isocyanate, 95+%, Thermo Scientific™

Product Code. 10580353
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
This item is not returnable. View return policy

Product Code. 10580353

Brand: Thermo Scientific Maybridge RF05240DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 58749-51-2
Molecular Formula C7H7NOS
Molecular Weight (g/mol) 153.199
InChI Key KAJZMNLREUEZSH-UHFFFAOYSA-N
PubChem CID 2782179
IUPAC Name 2-(2-isocyanatoethyl)thiophene
SMILES C1=CSC(=C1)CCN=C=O

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 2-2-isocyanatoethyl thiophene, 2-2-thienyl ethyl isocyanate, 2-thien-2-yl ethyl isocyanate, thiophene,2-2-isocyanatoethyl, 2-2-isocyanato-ethyl-thiophene, 2-2-thienyl ethanisocyanate, 2-2-isocyanatoethyl-thiophene, thiophene, 2-2-isocyanatoethyl, 2-thien-2-yl ethylisocyanate
Formula Weight 153.2
Percent Purity 95%
Chemical Name or Material 2-(2-Thienyl)ethyl isocyanate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.