missing translation for 'onlineSavingsMsg'
Learn More

2,3,3-Trimethylindolenine, 98%

Product Code. 10295090 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Packaging:
Glass bottle
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10295090 25 g Glass bottle 25g
10551771 100 g Glass bottle 100g
10561771 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10295090 Supplier Thermo Scientific Acros Supplier No. 140160250
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1640-39-7 | C11H13N | 159.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1640-39-7
Molecular Formula C11H13N
Molecular Weight (g/mol) 159.23
MDL Number MFCD00005724
InChI Key FLHJIAFUWHPJRT-UHFFFAOYSA-N
Synonym 2,3,3-trimethylindolenine, 2,3,3-trimethyl-3h-indole, 3h-indole, 2,3,3-trimethyl, ccris 6607, 2,3,3-trimethyl-indolenin, 2,3,3-trimethyl-3-hydroindole, pubchem7445, rarechem ah bs 0130, 2,3-trimethylindolenine, 2,3,3-trimethyl-indole
PubChem CID 15427
IUPAC Name 2,3,3-trimethylindole
SMILES CC1=NC2=CC=CC=C2C1(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 0.9900g/mL
Boiling Point 228°C to 229°C (744.0 mmHg)
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.545
Quantity 25 g
Beilstein 20, 324
Specific Gravity 0.99
Formula Weight 159.23
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 3, 3-Trimethylindolenine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2,3,3-Trimethylindolenine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.