missing translation for 'onlineSavingsMsg'
Learn More

2',3',4',5',6'-Pentafluoroacetophenone, 97%, ACROS Organics™

Product Code. 10286190 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
2.5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10286190 2.5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10286190 Supplier Thermo Scientific Acros Supplier No. 188310025
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 652-29-9
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.47g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C8H3F5O
Linear Formula C6F5COCH3
Refractive Index 1.4350 to 1.4370 (20°C, 589nm)
MDL Number MFCD00000296
Quantity 2.5 g
Synonym 2',3',4',5',6'-pentafluoroacetophenone, 1-perfluorophenyl ethanone, pentafluoroacetophenone, 1-pentafluorophenyl ethan-1-one, ethanone, 1-pentafluorophenyl, 2,3,4,5,6-pentafluoroacetophenone, 1-pentafluorphenyl ethanon, 1-acetyl-2,3,4,5,6-pentafluorobenzene, acmc-209npz, 1-pentafluorophenylethanone
InChI Key FBGHCYZBCMDEOX-UHFFFAOYSA-N
SMILES CC(=O)C1=C(F)C(F)=C(F)C(F)=C1F
IUPAC Name 1-(2,3,4,5,6-pentafluorophenyl)ethan-1-one
Molecular Weight (g/mol) 210.10
PubChem CID 69546
Formula Weight 210.1
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 2',3',4',5',6'-Pentafluoroacetophenone, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.