missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluorobenzenesulfonyl chloride, 98+%

Product Code. 11464120
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11464120

Brand: Thermo Scientific Alfa Aesar A17676.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 832-53-1 | C6ClF5O2S | 266.566 g/mol

Pentafluorobenzenesulfonyl chloride is used as derivatizing reagent for simultaneous determination of fluoxetine and p-trifluoromethylphenol, an O-dealkylated metabolite of fluoxetine in human liver microsomes. It is also used for electrophore labeling of small tyrosyl peptides for their analysis by gas chromatography with electron-capture detection. It is also employed as derivatizing reagent for amphetamine in determination of amphetamine in human plasma samples using gas chromatography with electron-capture detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluorobenzenesulfonyl chloride is used as derivatizing reagent for simultaneous determination of fluoxetine and p-trifluoromethylphenol, an O-dealkylated metabolite of fluoxetine in human liver microsomes. It is also used for electrophore labeling of small tyrosyl peptides for their analysis by gas chromatography with electron-capture detection. It is also employed as derivatizing reagent for amphetamine in determination of amphetamine in human plasma samples using gas chromatography with electron-capture detection.

Solubility
Soluble in water (reacts slowly).

Notes
Moisture Sensitive. Store away from oxidizing agents, active metals and bases, water/moisture. Keep the container tightly closed in a cool, dry place. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 832-53-1
Molecular Formula C6ClF5O2S
Molecular Weight (g/mol) 266.566
MDL Number MFCD00007427
InChI Key UOJCTEGNHXRPKO-UHFFFAOYSA-N
Synonym pentafluorobenzenesulfonyl chloride, 2,3,4,5,6-pentafluorobenzene-1-sulfonyl chloride, pentafluorobenzenesulphonyl chloride, benzenesulfonyl chloride, pentafluoro, pentafluorophenylsulfonyl chloride, 2,3,4,5,6-pentafluorobenzenesulfonyl choride, 2,3,4,5,6-pentafluoro-benzenesulfonyl chloride, chloro 2,3,4,5,6-pentafluorophenyl sulfone, pubchem5683, pentafluorobenzenesulfonylchloride
PubChem CID 70026
IUPAC Name 2,3,4,5,6-pentafluorobenzenesulfonyl chloride
SMILES C1(=C(C(=C(C(=C1F)F)S(=O)(=O)Cl)F)F)F

Specifications

Density 1.796
Boiling Point 210°C to 211°C
Flash Point 90°C (194°F)
Refractive Index 1.479
Quantity 1 g
UN Number UN3265
Beilstein 2142325
Sensitivity Moisture sensitive
Solubility Information Soluble in water (reacts slowly).
Formula Weight 266.57
Percent Purity ≥98%
Chemical Name or Material 2,3,4,5,6-Pentafluorobenzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.