missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose, 98%

Product Code. 15470597
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470597 100 g 100g
15450597 10 g 10g
15460597 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15470597 Supplier Thermo Scientific Chemicals Supplier No. J66537.22
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 20880-92-6 | C12H20O6 | 260.286 g/mol

2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose is used as chiral auxiliaries in Michael and Aldol addition reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose is used as chiral auxiliaries in Michael and Aldol addition reactions.

Solubility
Soluble in chloroform, ethyl acetate, methanol.

Notes
Protect from heat. Incompatible with heat and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20880-92-6
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.286
MDL Number MFCD00022183
InChI Key PSSHGMIAIUYOJF-OZFQHSNDSA-N
Synonym 2,3:4,5-bis-o-1,2 methylethylidene-beta-d-fruetopyranose
PubChem CID 45357248
IUPAC Name [(3aR,5aS,8aS)-2,2,7,7-tetramethyl-5,5a,8a,8b-tetrahydrodi[1,3]dioxolo[4,5-a
SMILES CC1(OC2COC3(C(C2O1)OC(O3)(C)C)CO)C

Specifications

Melting Point 93°C to 96°C
Color White
Quantity 100 g
Solubility Information Soluble in chloroform,ethyl acetate,methanol.
Optical Rotation −25.5° (c=1 in Chloroform)
Formula Weight 260.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.