missing translation for 'onlineSavingsMsg'
Learn More

2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose, TRC

Product Code. 30134459
Change view
Click to view available options
Quantity:
100 mg
1 g
2.5 g
Unit Size:
100mg
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134459 2.5 g 2.5g
30012379 1 g 1g
17938524 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134459 Supplier Toronto Research Chemicals Supplier No. TRCD4564002.5G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20880-92-6
Molecular Formula C12 H20 O6
Molecular Weight (g/mol) 260.28
Synonym 2,3:4,5-Bis-O-(1-methylethylidene)-beta-D-fructopyranose, β-D-Fructopyranose, 2,3:4,5-bis-O-(1-methylethylidene)-, Fructopyranose, 2,3:4,5-di-O-isopropylidene-, β-D- (6CI,7CI,8CI), 2,3:4,5-Bis-O-(1-methylethylidene)-β-D-fructopyranose, 5H-Bis[1,3]dioxolo[4,5-b:4',5'-d]pyran, β-D-fructopyranose deriv., 2,3:4,5-Di-O-isopropylidene-β-D-fructopyranose, D-Fructopyranose diacetonide, NSC 407023
IUPAC Name [(3aS,5aR,8aR,8bS)-2,2,7,7-tetramethyl-5,5a,8a,8b-tetrahydrodi[1,3]dioxolo[5,4-b:3',5'-e]pyran-3a-yl]methanol
SMILES CC1(C)O[C@@H]2CO[C@@]3(CO)OC(C)(C)O[C@H]3[C@@H]2O1

Spécification

Quantity 2.5 g
InChI Formula InChI=1S/C12H20O6/c1-10(2)15-7-5-14-12(6-13)9(8(7)16-10)17-11(3,4)18-12/h7-9,13H,5-6H2,1-4H3/t7-,8-,9+,12+/m1/s1
Formula Weight 260.126
Percent Purity >95%
Chemical Name or Material 2,3:4,5-Bis-O-(1-methylethylidene)-beta-D-fructopyranose
Grade HPLC
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.