missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5-Tetra-O-benzoyl-Alpha-D-glucopyranosyl bromide, TRC

Product Code. 30409674
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409674 50 mg -
30415379 100 mg -
30409841 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30409674 Supplier Toronto Research Chemicals Supplier No. TRCT28428850MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14218-11-2
Molecular Formula C34H27O9Br
Molecular Weight (g/mol) 659.48
Synonym Glucopyranosyl bromide, tetrabenzoate (7CI), Glucopyranosyl bromide, tetrabenzoate, (8CI), alpha-D-Glucopyranosyl bromide, tetrabenzoate (9CI), 2,3,4,5-Tetra-O-benzoyl-alpha-D-glucopyranosyl bromide, 2,3,4,5-Tetra-O-benzoyl-a-D-glucopyranosyl bromide, 2,3,4,6-Tetra-O-benzoyl-alpha-D-glucopyranosyl bromide, Tetra-O-benzoyl-alpha-D-glucopyranosyl bromide
IUPAC Name [(2R,3R,4S,5R,6R)-3,4,5-tribenzoyloxy-6-bromooxan-2-yl]methyl benzoate
SMILES Br[C@H]1O[C@H](COC(=O)c2ccccc2)[C@@H](OC(=O)c3ccccc3)[C@H](OC(=O)c4ccccc4)[C@H]1OC(=O)c5ccccc5

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C34H27BrO9/c35-30-29(44-34(39)25-19-11-4-12-20-25)28(43-33(38)24-17-9-3-10-18-24)27(42-32(37)23-15-7-2-8-16-23)26(41-30)21-40-31(36)22-13-5-1-6-14-22/h1-20,26-30H,21H2/t26-,27-,28+,29-,30+/m1/s1
Formula Weight 658.0838
Chemical Name or Material 2,3,4,5-Tetra-O-benzoyl-alpha-D-glucopyranosyl bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.