missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5-Tetrabromobenzoic Acid-13C6, TRC

Product Code. 30410761
Change view
Click to view available options
Quantity:
0.25 mg
2.5 mg
25 mg
Unit Size:
0.25mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30410761 2.5 mg 2.5mg
30426582 0.25 mg 0.25mg
30418637 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30410761 Supplier Toronto Research Chemicals Supplier No. TRCT2911422.5MG
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3,4,5-Tetrabromobenzoic Acid-13C6
Molecular Formula C13C6H2Br4O2
Quantity 2.5 mg
InChI Formula InChI=1S/C7H2Br4O2/c8-3-1-2(7(12)13)4(9)6(11)5(3)10/h1H,(H,12,13)/i1+1,2+1,3+1,4+1,5+1,6+1
SMILES Br[13C]1=[13C](Br)[13C](Br)=[13C](Br)[13C]([H])=[13C]1C(O)=O
Molecular Weight (g/mol) 443.66
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.