missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2,3,4,6-Tetra-O-benzyl-D-glucopyranose, 98+%

Product Code. 11357519
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357519 5 g 5g
11347519 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357519 Supplier Thermo Scientific Chemicals Supplier No. H27888.06
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 4132-28-9 | C34H36O6 | 540.656 g/mol

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in chloroform

Notes
Store -20°C. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4132-28-9
Molecular Formula C34H36O6
Molecular Weight (g/mol) 540.656
MDL Number MFCD00066004
InChI Key OGOMAWHSXRDAKZ-BKJHVTENSA-N
Synonym 2,3,4,6-tetra-o-benzyl-d-glucopyranose, 3r,4s,5r,6r-3,4,5-tris benzyloxy-6-benzyloxy methyl tetrahydro-2h-pyran-2-ol, tetrabenzylglucose, 3r,4s,5r,6r-3,4,5-tris benzyloxy-6-benzyloxy methyl oxan-2-ol, d-glucopyranose, 2,3,4,6-tetrakis-o-phenylmethyl, ksc235o0p, 2,3,4,6-tetra-o-benzylglucopyranose, 2,3,4,6-tetra-o-benzyl glucopyranose, 2,3,4,6-tetra-o-benzyl-glucopyranose, 2,3,4,6-terta-o-benzyl-d-glucopyranose
PubChem CID 10940502
IUPAC Name (3R,4S,5R,6R)-3,4,5-tris(phenylmethoxy)-6-(phenylmethoxymethyl)oxan-2-ol
SMILES C1=CC=C(C=C1)COCC2C(C(C(C(O2)O)OCC3=CC=CC=C3)OCC4=CC=CC=C4)OCC5=CC=CC=C5

Specifications

Melting Point 150°C to 155°C
Quantity 5 g
Solubility Information Soluble in chloroform
Formula Weight 540.65
Percent Purity ≥98%
Chemical Name or Material 2,3,4,6-Tetra-O-benzyl-D-glucopyranose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.