missing translation for 'onlineSavingsMsg'
Learn More

2,3',4'-Trichloroacetophenone

Product Code. 11477076
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11477076 25 g 25g
11467076 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11477076 Supplier Thermo Scientific Alfa Aesar Supplier No. B25397.14
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 42981-08-8 | C8H5Cl3O | 223.48 g/mol

2,3',4'-Trichloroacetophenone is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3′,4′-Trichloroacetophenone is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42981-08-8
Molecular Formula C8H5Cl3O
Molecular Weight (g/mol) 223.48
MDL Number MFCD03970382
InChI Key BYTZWANJVUAPNO-UHFFFAOYSA-N
Synonym 2,3',4'-trichloroacetophenone, 2-chloro-1-3,4-dichlorophenyl ethanone, 3,4-dichlorophenacyl chloride, 2-chloro-1-3,4-dichloro-phenyl-ethanone, 2-chloro-1-3,4-dichlorophenyl ethan-1-one, 2,3 ,4-trichloroacetophenone, pubchem20021, acmc-1bn8g, alpha,3',4'-trichloroacetophenone, 2-chloranyl-1-3,4-dichlorophenyl ethanone
PubChem CID 316188
SMILES ClCC(=O)C1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point 43°C to 45°C
Boiling Point 171°C to 173°C (15 mmHg)
Quantity 25 g
UN Number UN1759
Solubility Information Soluble in methanol.
IUPAC Name 2-chloro-1-(3,4-dichlorophenyl)ethan-1-one
Formula Weight 223.49
Chemical Name or Material 2,3',4'-Trichloroacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.