missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trihydroxyacetophenone, TRC

Product Code. 30454759
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454759 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30454759 Supplier Toronto Research Chemicals Supplier No. TRCT794905100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3,4-Trihydroxyacetophenone
CAS 528-21-2
Molecular Formula C8H8O4
Quantity 100 mg
Synonym 1-(2,3,4-Trihydroxyphenyl)-ethanone, 2',3',4'-Trihydroxy-acetophenone, Gallacetophenone, 1-(2,3,4-Trihydroxyphenyl)ethanone, 2',3',4'-Trihydroxyacetophenone, 4-Acetylpyrogallol, Alizarin Yellow C, Alizarine Yellow C, C.I. 57000, NSC 66553
InChI Formula InChI=1S/C8H8O4/c1-4(9)5-2-3-6(10)8(12)7(5)11/h2-3,10-12H,1H3
SMILES CC(=O)c1ccc(O)c(O)c1O
IUPAC Name 1-(2,3,4-trihydroxyphenyl)ethanone
Molecular Weight (g/mol) 168.15
Formula Weight 168.0423
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.