missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trihydroxybenzaldehyde, 98%

Product Code. 10462795 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10462795 100 g Glass bottle 100g
10324702 5 g Glass bottle 5g
10187700 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10462795 Supplier Thermo Scientific Acros Supplier No. 421791000
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2144-08-3 | C7H6O4 | 154.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2144-08-3
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.12
MDL Number MFCD00003325
InChI Key CRPNQSVBEWWHIJ-UHFFFAOYSA-N
Synonym pyrogallol-4-carboxaldehyde, benzaldehyde, 2,3,4-trihydroxy, pyrogallolaldehyd, pubchem8260, 2,4-trihydroxybenzaldehyde, dsstox_cid_31473, dsstox_rid_97358, dsstox_gsid_57684, ksc490m2j, benzaldehyde,3,4-trihydroxy
PubChem CID 75064
IUPAC Name 2,3,4-trihydroxybenzaldehyde
SMILES C1=CC(=C(C(=C1C=O)O)O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 159.0°C to 162.0°C
Color Beige-Brown to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 100 g
Formula Weight 154.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2, 3, 4-Trihydroxybenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2,3,4-Trihydroxybenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.