missing translation for 'onlineSavingsMsg'
Learn More

2-(3,5-Dichloro-4-oxopyridin-1(4H)-yl)acetic acid, 98%, BLD Pharm™

Product Code. 31096699
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31096699 100 g
31098708 25 g
31103868 5 g
31096518 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31096699 Supplier BLD Pharm Supplier No. BD10340100g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 56187-37-2
Molecular Formula C7H5Cl2NO3
MDL Number MFCD02180547
Quantity 100 g
SMILES O=C(O)CN(C=C1Cl)C=C(Cl)C1=O
Molecular Weight (g/mol) 222.03
Percent Purity 98%
Chemical Name or Material 2-(3,5-Dichloro-4-oxopyridin-1(4H)-yl)acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.