missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Tri-O-benzyl-beta-D-arabinofuranose

Product Code. 15437539
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15437539

Brand: Thermo Scientific Alfa Aesar J66252.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 89361-52-4 | C26H28O5 | 420.505 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89361-52-4
Molecular Formula C26H28O5
Molecular Weight (g/mol) 420.505
MDL Number MFCD01864312
InChI Key NAQUAXSCBJPECG-VEYUFSJPSA-N
Synonym 2,3,5-tri-o-benzyl-b-d-ribofuranose, 2,3,5-tri-o-benzyl-beta-d-ribofuranose, 2r,3r,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl oxolan-2-ol, 2-o,3-o,5-o-tribenzyl-beta-d-ribofuranose, 2r,3r,4r,5r-3,4-bis phenylmethoxy-5-phenylmethoxymethyl, 2r,3r,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl tetrahydrofuran-2-ol
PubChem CID 57369909
IUPAC Name (2R,3R,4R,5R)-3,4-bis(phenylmethoxy)-5-(phenylmethoxymethyl)oxolan-2-ol
SMILES C1=CC=C(C=C1)COCC2C(C(C(O2)O)OCC3=CC=CC=C3)OCC4=CC=CC=C4

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in chloroform
Optical Rotation +43° (c=2 in Dioxane)
Formula Weight 420.5
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,3,5-Tri-O-benzyl-beta-D-ribofuranose

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.