missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Triphenyl-2H-tetrazolium Chloride, TRC

Product Code. 30454783
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454783 1 g -
30456373 5 g 5g
30453975 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30454783 Supplier Toronto Research Chemicals Supplier No. TRCT8091001G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-96-4
Molecular Formula C19H15ClN4
Molecular Weight (g/mol) 334.8
Synonym 1,3,5-Triphenyl-2H-tetrazolium Chloride, 1,3,5-Triphenyltetrazolium Chloride, 2,3,5-Triphenyltetrazolium Chloride, PTB, RT, Red Tetrazolium, TPTZ, TT, TTC, TTC (dye), Tetrazolium Chloride, Tetrazolium Red, Triphenyltetrazolium Chloride, Urocheck, Uroscreen, Vitastain
SMILES [N+]1(C2=CC=CC=C2)=NC(C3=CC=CC=C3)=NN1C4=CC=CC=C4.[Cl-]

Specifications

Quantity 1 g
Formula Weight 334.0985
Chemical Name or Material 2,3,5-Triphenyl-2H-tetrazolium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.