missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Triphenyltetrazolium, Technical, Fisher Chemical™

Product Code. 12807192 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12807192 100 g 100g
12615296 10 g 10g
12625296 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12807192 Supplier Fisher Chemical Supplier No. T/3700/48
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

C19H15ClN4, CAS Number-298-96-4, 2,3,5-triphenyltetrazolium chloride, triphenyltetrazolium chloride, 2,3,5-triphenyl-2h-tetrazolium chloride, tetrazolium chloride, red tetrazolium, tetrazolium red, tetrazolium chloride, uroscreen, urocheck, vitastain, 25g, CHEBI:78019, White, 334.81g/mol | CAS: 298-96-4 | C19H15ClN4 | 334.81 g/mol

C19H15ClN4, CAS Number-298-96-4, 2,3,5-triphenyltetrazolium chloride, triphenyltetrazolium chloride, 2,3,5-triphenyl-2h-tetrazolium chloride, tetrazolium chloride, red tetrazolium, tetrazolium red, tetrazolium chloride, uroscreen, urocheck, vitastain, 25g, CHEBI:78019, White, 334.81g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-96-4
Molecular Formula C19H15ClN4
Molecular Weight (g/mol) 334.81
MDL Number MFCD00011963
InChI Key PKDBCJSWQUOKDO-UHFFFAOYSA-M
Synonym 2,3,5-triphenyltetrazolium chloride, tetrazolium red, uroscreen, red tetrazolium, urocheck, vitastain, tetrazolium chloride, triphenyltetrazolium chloride, 2,3,5-triphenyl-2h-tetrazolium chloride, tetrazolium chloride
PubChem CID 9283
ChEBI CHEBI:78019
IUPAC Name 2,3,5-triphenyltetrazol-2-ium;chloride
SMILES [Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 235°C
Color White
Packaging Plastic powder jar
Quantity 100 g
Formula Weight 334.81g/mol
Physical Form Solid
compliance-icons
Product Identifier
  • 2,3,5-Triphenyl-2H-tetrazolium chloride
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.