missing translation for 'onlineSavingsMsg'
Learn More

2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-lactosyl bromide

Product Code. 15470567
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15470567 5 g 5g
15460567 1 g 1g
15480567 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15470567 Supplier Thermo Scientific Alfa Aesar Supplier No. J66385.06
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 4753-07-5 | C26H35BrO17 | 699.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4753-07-5
Molecular Formula C26H35BrO17
Molecular Weight (g/mol) 699.45
MDL Number MFCD00153593
InChI Key NLFHLQWXGDPOME-UHFFFAOYNA-N
Synonym bromo heptaacetyl-d-lactoside, stabilized with calcium carbonate, bromo heptaacetyl-d-lactoside, 2,3,6,2 ,3 ,4 ,6-hepta-o-acetyl-a-d-cellobiosyl bromide, 2,2',3,3',4',6,6'-hepta-o-acetyl-alpha-d-lactosyl bromide, 2,3,6,2',3',4',6'-hepta-o-acetyl-1-bromo-1-deoxy-alpha-d-lactopyranose min, 2r,3s,4s,5r,6s-3,4,5-tris acetyloxy-6-2r,3r,4s,5r,6r-4,5-bis acetyloxy-2-acetyloxy methyl-6-bromooxan-3-yl oxy oxan-2-yl methyl acetate, 2-o,3-o,6-o-triacetyl-4-o-2-o,3-o,4-o,6-o-tetraacetyl-beta-d-galactopyranosyl-alpha-d-glucopyranosyl bromide, acetobromo-, a-d-lactoside, bromo 4-o-2 ,3,4,6-tetra-o-acetyl-, a-d-galactopyranosyl-2,3,6-tri-o-acetyl-, a-d-glucopyranoside,
PubChem CID 11136238
IUPAC Name [(2R,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-[(2R,3R,4S,5R,6R)-4,5-diacetyloxy-2-(acetyloxymethyl)-6-bromooxan-3-yl]oxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC2C(OC(C(C2OC(=O)C)OC(=O)C)Br)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Color White
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in chloroform
Formula Weight 699.45
Physical Form Powder
Chemical Name or Material 2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-lactosyl bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.