missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,3,6-Trifluoropyridine-4-carboxylic acid, tech. 90%

Product Code. 11314476
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11314476 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11314476 Supplier Alfa Aesar™ Supplier No. L20051.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 675602-92-3
Molecular Formula C6H2F3NO2
Molecular Weight (g/mol) 177.082
MDL Number MFCD03840204
InChI Key RXNORRUCMWSIQZ-UHFFFAOYSA-N
Synonym 2,3,6-trifluoroisonicotinic acid, 2, 3, 6-trifluoroisonicotinic acid, 2,3,6-trifluoro-4-pyridinecarboxylic acid, 4-pyridinecarboxylicacid,2,3,6-trifluoro, 4-pyridinecarboxylic acid, 2,3,6-trifluoro, 2,3,6-tris fluoranyl pyridine-4-carboxylic acid, 2,3,6-trifluoropyridine-4-carboxylic acid
PubChem CID 2782837
IUPAC Name 2,3,6-trifluoropyridine-4-carboxylic acid
SMILES C1=C(C(=C(N=C1F)F)F)C(=O)O

Specifications

Melting Point 118°C to 121°C
Quantity 1 g
Formula Weight 177.08
Percent Purity 90%
Grade Technical
Chemical Name or Material 2,3,6-Trifluoropyridine-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.