missing translation for 'onlineSavingsMsg'
Learn More

2,3-Bis(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Product Code. 11378746
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11378746

Brand: Thermo Scientific Alfa Aesar L19445.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1644-68-4
Molecular Formula C7H3F6N
Molecular Weight (g/mol) 215.098
MDL Number MFCD03093763
InChI Key RRNXYHYDSDAOFW-UHFFFAOYSA-N
Synonym 2,3-bis trifluoromethyl pyridine, pyridine, 2,3-bis trifluoromethyl, pubchem7694, acmc-209dra, bis trifluoromethyl pyridine, 2,3-bis trifluoromethyl pyridine #, pyridine,2,3-bis trifluoromethyl
PubChem CID 589865
IUPAC Name 2,3-bis(trifluoromethyl)pyridine
SMILES C1=CC(=C(N=C1)C(F)(F)F)C(F)(F)F

Specifications

Melting Point -2°C
Boiling Point 85°C (18 mmHg)
Flash Point 70°C (158°F)
Quantity 250 mg
UN Number UN2810
Beilstein 1622028
Formula Weight 215.1
Percent Purity 97%
Chemical Name or Material 2,3-Bis(trifluoromethyl)pyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.