missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Bromopropoxy)tetrahydro-2H-pyran, 98%, stabilized with potassium carbonate, Thermo Scientific Chemicals

Product Code. 10705915 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10705915 5 g 5g
10481654 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10705915 Supplier Thermo Scientific Acros Supplier No. 399730050
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33821-94-2
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.1
InChI Key HJNHUFQGDJLQRS-UHFFFAOYSA-N
Synonym 2-3-bromopropoxy tetrahydro-2h-pyran, 2-3-bromopropoxy oxane, 2-3-bromopropoxy-tetrahydro-2h-pyran, 2-3-bromopropoxy tetrahydropyran, 1-bromo-3-tetrahydropyranyloxypropane, 2-3-bromo-propoxy-tetrahydro-pyran, 2h-pyran, 2-3-bromopropoxy tetrahydro, acmc-1cjr6, 1-bromo-3-2-oxanyloxy propane
PubChem CID 2777988
IUPAC Name 2-(3-bromopropoxy)oxane
SMILES C1CCOC(C1)OCCCBr

Specifications

Density 1.3140g/mL
Boiling Point 64.5°C (0.8 mmHg)
Flash Point 106°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 5 g
Specific Gravity 1.314
Formula Weight 223.1
Percent Purity 98%
Chemical Name or Material 2-(3-Bromopropoxy)tetrahydro-2H-pyran, Stabilized with potassium carbonate
compliance-icons
Product Identifier
  • 2-(3-Bromopropoxy)tetrahydro-2H-pyran, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.