missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Butanediol-d8 (Mixture of Diastereomers), TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | 2,3-Butanediol-d8 |
| CAS | 347841-77-4 |
| Molecular Formula | C4 D8 H2 O2 |
| Quantity | 10 mg |
| Synonym | 2,3-Butane-1,1,1,2,3,4,4,4-d8-diol (9CI), 2,3-Butanediol-d8, 2,3-Butanediol-D8, 2,3-Butanediol-1,1,1,2,3,4,4,4-d8, 2,3-Butane-d8-diol |
| InChI Formula | InChI=1S/C4H10O2/c1-3(5)4(2)6/h3-6H,1-2H3/i1D3,2D3,3D,4D |
| SMILES | [2H]C([2H])([2H])C([2H])(O)C([2H])(O)C([2H])([2H])[2H] |
| IUPAC Name | 1,1,1,2,3,4,4,4-octadeuteriobutane-2,3-diol |
| Molecular Weight (g/mol) | 98.17 |
| Formula Weight | 98.118 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction