missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Butynyloxy)tetrahydro-2H-pyran, 97%

Product Code. 10617040 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10617040 5 g Glass bottle 5g
10547600 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10617040 Supplier Thermo Scientific Acros Supplier No. 428210050
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 40365-61-5 | C9H14O2 | 154.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 40365-61-5
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.21
MDL Number MFCD00012352
InChI Key ZQZSNKJFOFAJQX-UHFFFAOYSA-N
Synonym 2-but-3-yn-1-yloxy tetrahydro-2h-pyran, 2-3-butynyloxy tetrahydro-2h-pyran, 2h-pyran, 2-3-butynyloxy tetrahydro, 2-3-butynyloxy tetrahydro-2 h-pyran, 1-tetrahydropyran-2-yloxy-3-butyne, 2-but-3-ynoxytetrahydropyran, tetrahydropyranyloxy-3-butyne, 2-but-3-yn-1-yloxy oxane, 2-3-butynyloxy tetrahydropyran, 4-tetrahydropyranyl oxy butyne
PubChem CID 142440
IUPAC Name 2-but-3-ynoxyoxane
SMILES C#CCCOC1CCCCO1

Spezifikation

CAS Min % 96.0
CAS Max % 100.0
Density 0.9840g/mL
Boiling Point 92.0°C to 95.0°C (18.0 mmHg)
Flash Point 73°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 0.984
Formula Weight 154.21
Percent Purity 97%
Chemical Name or Material 2-(3-Butynyloxy)tetrahydro-2H-pyran
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • 2-(3-Butynyloxy)tetrahydro-2H-pyran
Signalwort
  • Warning
Gefahrenkategorie
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.